Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Methylenebis(6-cyclohexyl-p-cresol) 97.0+%, TCI America™
SDP

Catalog No. M189625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M189625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M189625G Supplier TCI America Supplier No. M189625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4066-02-8
Melting Point 134°C
Color White-Yellow
Molecular Formula C27H36O2
Quantity 25 g
Synonym 2,2′C-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
InChI Key AKNMPWVTPUHKCG-UHFFFAOYSA-N
SMILES CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O
IUPAC Name 2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Molecular Weight (g/mol) 392.58
PubChem CID 77681
Formula Weight 392.58
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(6-cyclohexyl-p-cresol)
Show More Show Less
RTECSNumber SM0335000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.