accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4067-16-7

IUPAC Name: 3,6,9,12-tetraazatetradecane-1,14-diamine
Synonyms: pentaethylenehexamine
Molecular Weight (g/mol): 232.38
Molecular Formula: C10H28N6
InChi Key: LSHROXHEILXKHM-UHFFFAOYSA-N
SMILES: NCCNCCNCCNCCNCCN
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)