Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaethylenehexamine

Catalog No. 292753250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-275-3250ML 250 mL
29-275-31L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-275-3250ML Supplier Sigma Aldrich Supplier No. 292753250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5.pentaethylenehexamine has been used in the preparation of:
  • Polyisobutylene succinimide (pibsi) dispersants, used as engine oil-additives
  • Poly(glycidyl methacrylate) (p(gma)) homopolymers
  • Poly(amidoamine)s

Specifications

CAS 4067-16-7
Density 0.95 g/mL (at 25°C (literature))
Molecular Formula C10H28N6
Linear Formula NH2CH2CH2NH(CH2CH2NH)3 CH2CH2NH2
Refractive Index n20/D 1.5096 (literature)
MDL Number MFCD00008167
Quantity 250 mL
Synonym 3,6,9,12-Tetraazatetradecane-1,14-diamine
Molecular Weight (g/mol) 232.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.