accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H2N

CAS RN 40691-66-5

IUPAC Name: hydrogen 2,2-diphenylpropan-1-amine chloride
Synonyms: hydrogen 2,2-diphenylpropylamine chloride
Molecular Weight (g/mol): 247.77
Molecular Formula: C15H18ClN
InChi Key: AASCJSPDUDWGGQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(CN)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results