Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylpropylamine Hydrochloride 95%, Thermo Scientific™

Catalog No. AC182130050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182130050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182130050 Supplier Thermo Scientific Chemicals Supplier No. 182130050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40691-66-5
Molecular Formula C15H18ClN
Molecular Weight (g/mol) 247.766
InChI Key AASCJSPDUDWGGQ-UHFFFAOYSA-N
Synonym 2,2-diphenylpropylamine hydrochloride, 2,2-diphenylpropan-1-amine hydrochloride, beta-methyl-beta-phenylphenethylamine hydrochloride, acmc-20aojd, diphenylpropylamine hci, beta-methyl-beta-phenylphenethylamine hcl
PubChem CID 3084798
IUPAC Name 2,2-diphenylpropan-1-amine;hydrochloride
SMILES CC(CN)(C1=CC=CC=C1)C2=CC=CC=C2.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Linear Formula CH3C(C6H5)2CH2NH2.HCl
Quantity 5 g
Formula Weight 247.77
Percent Purity ≥94%
Physical Form Granular Powder
Chemical Name or Material 2, 2-Diphenylpropylamine hydrochloride, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.