accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40738-83-8

IUPAC Name: 5-iodo-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-iodo-1,3-dimethylpyrimidine-2,4-dione
Molecular Weight (g/mol): 266.04
Molecular Formula: C6H7IN2O2
InChi Key: XUUVRIXNFFFPCM-UHFFFAOYSA-N
SMILES: CN1C=C(I)C(=O)N(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results