accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40787-48-2

IUPAC Name: 1,4-dibromo-2,5-diethylbenzene
Synonyms: 1,4-dibromo-2,5-diethylbenzene
Molecular Weight (g/mol): 292.01
Molecular Formula: C10H12Br2
InChi Key: DNHRBCFMFYOUKM-UHFFFAOYSA-N
SMILES: CCC1=CC(Br)=C(CC)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results