Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2,5-diethylbenzene 98.0+%, TCI America™
SDP

Catalog No. D28075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28075G 5 g
D28071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28075G Supplier TCI America Supplier No. D28075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40787-48-2
Molecular Formula C10H12Br2
Molecular Weight (g/mol) 292.01
MDL Number MFCD04038416
InChI Key DNHRBCFMFYOUKM-UHFFFAOYSA-N
Synonym 2,5-Diethyl-1,4-dibromobenzene
PubChem CID 21501836
IUPAC Name 1,4-dibromo-2,5-diethylbenzene
SMILES CCC1=CC(Br)=C(CC)C=C1Br

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 5 g
Formula Weight 292.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-diethylbenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.