accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSHCH3O

CAS RN 40789-98-8

IUPAC Name: 3-sulfanylbutan-2-one
Synonyms: 2-butanone, 3-mercapto-
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: XLMPYCGSRHSSSX-UHFFFAOYNA-N
SMILES: CC(S)C(C)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)