Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-2-butanone 96.0+%, TCI America™
SDP

Catalog No. M184425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M184425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M184425G Supplier TCI America Supplier No. M184425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40789-98-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.17
MDL Number MFCD00085206
InChI Key XLMPYCGSRHSSSX-UHFFFAOYNA-N
Synonym 3-mercapto-2-butanone, 2-butanone, 3-mercapto, 2-mercapto-3-butanone, 3-mercaptobutan-2-one, fema no. 3298, mercapto-3 butanone-2 french, mercapto-3 butanone-2, acmc-209jgq, 4-01-00-03993 beilstein handbook reference
PubChem CID 38679
IUPAC Name 3-sulfanylbutan-2-one
SMILES CC(S)C(C)=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
UN Number 3336
Formula Weight 104.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-2-butanone
RTECSNumber EL9050000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.