accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOHOOOONH2

CAS RN 4079-64-5

IUPAC Name: 1,4-dibenzyl 2-aminobutanedioate; 4-methylbenzene-1-sulfonic acid
Synonyms: 1,4-dibenzyl 2-aminobutanedioate; para-toluene sulfonate
Molecular Weight (g/mol): 485.55
Molecular Formula: C25H27NO7S
InChi Key: HLMUYZYLPUHSNV-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)