accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOHNN

CAS RN 40828-54-4

IUPAC Name: 1H-1,3-benzodiazole-2-sulfonate
Synonyms: 1H-1,3-benzodiazole-2-sulfonate
Molecular Weight (g/mol): 197.19
Molecular Formula: C7H5N2O3S
InChi Key: GWXQTTKUYBEZBP-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results