Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Benzimidazole-2-sulfonic Acid 98%, Thermo Scientific™

Catalog No. AC276320050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC276320050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276320050 Supplier Thermo Scientific Chemicals Supplier No. 276320050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40828-54-4
Molecular Formula C7H6N2O3S
Molecular Weight (g/mol) 198.196
InChI Key GWXQTTKUYBEZBP-UHFFFAOYSA-N
Synonym 1h-benzoimidazole-2-sulfonic acid, 1h-benzo d imidazole-2-sulfonic acid, benzimidazole-2-sulfonic acid, 1h-1,3-benzodiazole-2-sulfonic acid, 2-benzimidazolesulfonic acid, cambridge id 5140822, 1h-benzimidazolesulfonic acid, 1h-benzimidazole-2-sulfonicacid, 1h-benzimidazole-2-sulfonic acid, 9ci
PubChem CID 1572160
IUPAC Name 1H-benzimidazole-2-sulfonic acid
SMILES C1=CC=C2C(=C1)NC(=N2)S(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 198.2
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Benzimidazole-2-sulfonic acid, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.