accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4083-64-1

IUPAC Name: 4-methylbenzene-1-sulfonyl isocyanate
Synonyms: 4-methylbenzenesulfonyl isocyanate
Molecular Weight (g/mol): 197.21
Molecular Formula: C8H7NO3S
InChi Key: VLJQDHDVZJXNQL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N=C=O
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (7)
Quantity 
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (7)