Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich p-Toluenesulfonyl isocyanate

Catalog No. 189278500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-927-8500G 500 g
18-927-825G 25 g
18-927-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-927-8500G Supplier Sigma Aldrich Supplier No. 189278500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • P-toluenesulfonyl isocyanate undergoes palladium-catalyzed bis-allylation reaction with allylstannanes and allyl chlorides.p-toluenesulfonyl isocyanate has been used as :
  • Derivatization reagent in determination of 3-α-hydroxy tibolone in human plasma by LC-ms/ms
  • Derivatization reagent in simultaneous quantitative analysis of diethylene glycol and propylene glycol in pharmaceutical products by HPLC
  • As reagent in the preparation of acetylated syn-1,2-diols, oxazolidin-2-ones, 2,3-diamino acids and n-tosylcarbonamides

Specifications

CAS 4083-64-1
Density 1.291 g/mL (at 25°C (literature))
Boiling Point 144°C (10 mmHg), 275°C to 278°C (lit.)
Molecular Formula C8H7NO3S
Refractive Index n20/D 1.534 (literature)
Linear Formula CH3C6H4SO2NCO
MDL Number MFCD00002030
Quantity 500 g
Synonym Tosyl isocyanate
Molecular Weight (g/mol) 197.21
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.