CAS RN 40832-47-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-2-nitro-N,N-di-n-propylaniline, 97%, Thermo Scientific™
CAS: 40832-47-1 Synonym: (4-Chloro-2-nitrophenyl)di-n-propylamine