accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2H2NCl

CAS RN 40851-95-4

IUPAC Name: 6-chloropyridine-2,3-diamine
Synonyms: 6-chloropyridine-2,3-diamine
Molecular Weight (g/mol): 143.57
Molecular Formula: C5H6ClN3
InChi Key: QEIRYIILFUVXAM-UHFFFAOYSA-N
SMILES: NC1=C(N)N=C(Cl)C=C1
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)