Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diamino-6-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. D47651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47651G Supplier TCI America Supplier No. D47651G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity heterocyclic building block for chemical synthesis.

Chemical Identifiers

CAS 40851-95-4
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.57
MDL Number MFCD00209966
InChI Key QEIRYIILFUVXAM-UHFFFAOYSA-N
Synonym 2,3-diamino-6-chloropyridine, 6-chloro-2,3-diaminopyridine, 2,3-pyridinediamine,6-chloro, 2,3-pyridinediamine, 6-chloro, 6-chloro-pyridine-2,3-diamine, 6-chloro-2,3-pyridinediamine, pubchem19958, 6-chloro 2,3-pyridindiamine, 2,3-diamino-6-chloro-pyridine
PubChem CID 10630615
IUPAC Name 6-chloropyridine-2,3-diamine
SMILES NC1=C(N)N=C(Cl)C=C1

Specifications

Melting Point 134°C
Color Red-White
Quantity 1 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diamino-6-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.