CAS RN 409-21-2
Silicon carbide powder, medium, 120 grit
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide powder, superfine, 600 grit
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide whiskers, 99% (metals basis), Thermo Scientific Chemicals
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilanylium SMILES: [C-]#[Si+]
Silicon carbide, alpha-phase, 99.8% (metals basis)
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide, 400 grinding compound, 2oz (57g)
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilylium SMILES: [C-]#[Si+]
Silicon carbide, beta-phase, nanopowder, 95% min
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide powder, fine, 320 grit
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilanylium SMILES: [C-]#[Si+]
Silicon carbide powder, coarse, 46 grit
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide, 99% (metals basis)
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilanylium SMILES: [C-]#[Si+]
Silicon carbide, beta-phase, 99% (metals basis)
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilylium SMILES: [C-]#[Si+]
Silicon carbide, beta-phase, nanopowder
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 SMILES: [C-]#[Si+]
Silicon carbide, 600 grinding compound, 2oz (57g), Thermo Scientific™
CAS: 409-21-2 Molecular Formula: CSi Molecular Weight (g/mol): 40.10 MDL Number: MFCD00049531 InChI Key: HBMJWWWQQXIZIP-UHFFFAOYSA-N Synonym: silicon carbide,carborundum,silicon monocarbide,carborundeum,tokawhisker,betarundum,carbolon,nicalon,silundum,carbon silicide PubChem CID: 9863 ChEBI: CHEBI:29390 IUPAC Name: methanidylidynesilanylium SMILES: [C-]#[Si+]
Sigma Aldrich Silicon carbide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Silicon carbide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Silicon carbide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2