accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40912-11-6

IUPAC Name: methyl 4-(3-methoxy-3-oxopropyl)benzoate
Synonyms: methyl 4-(3-methoxy-3-oxopropyl)benzoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: MLKXCCLYRPKSBM-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=C(C=C1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results