Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(4-Methoxycarbonylphenyl)propionate 97.0+%, TCI America™
SDP

Catalog No. C10295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10295G 5 g
C102925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10295G Supplier TCI America Supplier No. C10295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40912-11-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00059305
InChI Key MLKXCCLYRPKSBM-UHFFFAOYSA-N
Synonym 3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester
PubChem CID 595759
IUPAC Name methyl 4-(3-methoxy-3-oxopropyl)benzoate
SMILES COC(=O)CCC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 33°C
Color White
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-(4-Methoxycarbonylphenyl)propionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.