accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40928-13-0

IUPAC Name: 7-chloro-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Synonyms: 7-chloro-1H-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol): 197.57
Molecular Formula: C8H4ClNO3
InChi Key: QRUPDIJQZCABTC-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=O)OC(=O)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results