Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroisatoic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C27901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27901G Supplier TCI America Supplier No. C27901G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40928-13-0
Molecular Formula C8H4ClNO3
Molecular Weight (g/mol) 197.57
MDL Number MFCD00090431
InChI Key QRUPDIJQZCABTC-UHFFFAOYSA-N
Synonym 7-Chloro-1,2-dihydro-4H-3,1-benzoxazine-2,4-dione
PubChem CID 329105
IUPAC Name 7-chloro-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
SMILES ClC1=CC=C2C(=O)OC(=O)NC2=C1

Specifications

Melting Point 253°C
Color Yellow
Quantity 1 g
Formula Weight 197.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloroisatoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.