accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 4096-20-2

IUPAC Name: 1-phenylpiperidine
Synonyms: N-phenylpiperidine
Molecular Weight (g/mol): 161.25
Molecular Formula: C11H15N
InChi Key: LLSKXGRDUPMXLC-UHFFFAOYSA-N
SMILES: C1CCN(CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results