Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenylpiperidine 98.0+%, TCI America™
SDP

Catalog No. P21591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21591G Supplier TCI America Supplier No. P21591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4096-20-2
Color Yellow
Boiling Point 258°C
Molecular Formula C11H15N
Quantity 1 g
MDL Number MFCD00129777
InChI Key LLSKXGRDUPMXLC-UHFFFAOYSA-N
SMILES C1CCN(CC1)C2=CC=CC=C2
IUPAC Name 1-phenylpiperidine
Molecular Weight (g/mol) 161.248
PubChem CID 20038
Formula Weight 161.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Phenylpiperidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.