accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2NNH2

CAS RN 4097-89-6

IUPAC Name: tris(2-aminoethyl)amine
Synonyms: Tren β,β',β''-triaminotriethylamine 2,2',2''-triaminotriethylamine triaminotriethylamine tren compound tren HP tren
Molecular Weight (g/mol): 146.24
Molecular Formula: C6H18N4
InChi Key: MBYLVOKEDDQJDY-UHFFFAOYSA-N
SMILES: NCCN(CCN)CCN
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)