Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(2-aminoethyl)amine

Catalog No. 22563010ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-563-010ML 10 mL
22-563-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-563-010ML Supplier Sigma Aldrich Supplier No. 22563010ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2-aminoethyl)amine (TREN) is a water soluble tripodal ligand that is majorly used in co-ordination chemistry. It has three aminoethylgroups that attach with the surface atoms to provide a scaffold assembly.TREN can be grafted with multi-walled carbon nanotube (MWCNT) for use in solid phase extraction of metal ions for wastewater treatment based applications. It can also be used as a chelating agent in the surface treatment of silica nanoparticles which can further be used for a wide range of industrial applications.

Specifications

CAS 4097-89-6
Density 0.976 g/mL (at 20°C)
Boiling Point 114°C (15 mmHg)
Molecular Formula C6H18N4
Refractive Index n20/D 1.497 (literature)
Linear Formula (NH2CH2CH2)3 N
Quantity 10 mL
MDL Number MFCD00008177
Synonym 2,2 ,2 -Nitrilotriethylamine; 2,2 ,2 -Triaminotriethylamine; TAEA
Molecular Weight (g/mol) 146.23
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.