accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(R)O(R)(R)OH3CO(R)OH3CONOHNO

CAS RN 4105-38-8

IUPAC Name: [(2R,3R,4R,5R)-3,4-bis(acetyloxy)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl acetate
Synonyms: [(2R,3R,4R,5R)-3,4-bis(acetyloxy)-5-(2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol): 370.31
Molecular Formula: C15H18N2O9
InChi Key: AUFUWRKPQLGTGF-FMKGYKFTSA-N
SMILES: CC(=O)OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H]1OC(C)=O)N1C=CC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results