Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',5'-Tri-O-acetyluridine 98.0+%, TCI America™
SDP

Catalog No. T25495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25495G 5 g
T254925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25495G Supplier TCI America Supplier No. T25495G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4105-38-8
Molecular Formula C15H18N2O9
Molecular Weight (g/mol) 370.31
MDL Number MFCD00023795
InChI Key AUFUWRKPQLGTGF-FMKGYKFTSA-N
Synonym Uridine 2′C,3′C,5′C-Triacetate
PubChem CID 124300592
IUPAC Name [(2R,3R,4R,5R)-3,4-bis(acetyloxy)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H]1OC(C)=O)N1C=CC(=O)NC1=O

Specifications

Melting Point 129°C
Color White
Quantity 5 g
Formula Weight 370.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,5′-Tri-O-acetyluridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.