accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 4105-95-7

IUPAC Name: 4-(octadecyloxy)benzaldehyde
Synonyms: 4-(octadecyloxy)benzaldehyde
Molecular Weight (g/mol): 374.61
Molecular Formula: C25H42O2
InChi Key: AKTNFINCBVUXEH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results