Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Octadecyloxybenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. O02235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02235G Supplier TCI America Supplier No. O02235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4105-95-7
Molecular Formula C25H42O2
Molecular Weight (g/mol) 374.609
MDL Number MFCD00143357
InChI Key AKTNFINCBVUXEH-UHFFFAOYSA-N
PubChem CID 4433898
IUPAC Name 4-octadecoxybenzaldehyde
SMILES CCCCCCCCCCCCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 374.61
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Octadecyloxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.