accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41051-80-3

CH3NH3CSiOH3COH3COH3C

CAS RN 41051-80-3

IUPAC Name: diethyl[3-(trimethoxysilyl)propyl]amine
Synonyms: diethyl[3-(trimethoxysilyl)propyl]amine
Molecular Weight (g/mol): 235.4
Molecular Formula: C10H25NO3Si
InChi Key: ZLDHYRXZZNDOKU-UHFFFAOYSA-N
SMILES: CCN(CC)CCC[Si](OC)(OC)OC
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)