Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylzinc Chloride Solution

Catalog No. 41729750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-729-750ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 41-729-750ML Supplier Sigma Aldrich Supplier No. 41729750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylzinc chloride can be used in: Ni-catalyzed one-pot Negishi coupling with amino-heteroaryl chlorides.Cross-coupling with aryl fluorides in the presence of a nickel catalyst.

Specifications

CAS 41051-80-3
Density 0.95 g/mL at 25°C
Boiling Point 120°C (20 mmHg, lit.)
Molecular Formula C10H25NO3Si
Linear Formula (C2H5)2N(CH2)3Si(OCH3)3
MDL Number MFCD00039882
Quantity 50 mL
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.