accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOH

CAS RN 41059-02-3

IUPAC Name: 9-bromononanoic acid
Synonyms: 9-bromononanoic acid
Molecular Weight (g/mol): 237.14
Molecular Formula: C9H17BrO2
InChi Key: XEGRKZRPTBNSMN-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results