Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromononanoic Acid 97.0+%, TCI America™
SDP

Catalog No. B23235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23235G 5 g
B23231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23235G Supplier TCI America Supplier No. B23235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41059-02-3
Molecular Formula C9H17BrO2
Molecular Weight (g/mol) 237.137
MDL Number MFCD00046564
InChI Key XEGRKZRPTBNSMN-UHFFFAOYSA-N
PubChem CID 548221
IUPAC Name 9-bromononanoic acid
SMILES C(CCCCBr)CCCC(=O)O

Specifications

Melting Point 37°C
Boiling Point 174°C
Quantity 5 g
UN Number 3261
Formula Weight 237.14
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromononanoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.