accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 4107-62-4

IUPAC Name: methyl 3-cyanopropanoate
Synonyms: methyl 3-cyanopropanoate
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: BPSKURPOKFSLHJ-UHFFFAOYSA-N
SMILES: COC(=O)CCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results