Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Cyanopropionate 99.0+%, TCI America™
SDP

Catalog No. C079225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C079225ML Supplier TCI America Supplier No. C079225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4107-62-4
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD00058965
InChI Key BPSKURPOKFSLHJ-UHFFFAOYSA-N
Synonym 3-Cyanopropionic Acid Methyl Ester
PubChem CID 77749
IUPAC Name methyl 3-cyanopropanoate
SMILES COC(=O)CCC#N

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 mL
UN Number 3276
Formula Weight 113.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Cyanopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.