CAS RN 41123-44-8
IUPAC Name:
1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11-icosafluoroundecyl 2-methylprop-2-enoate
Synonyms:
1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11-icosafluoroundecyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
600.2
Molecular Formula:
C15H8F20O2
InChi Key:
KDUYSMPJTYKVBA-UHFFFAOYNA-N
SMILES:
CC(=C)C(=O)OC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CF
1
–
1
of
1
results
1
1H,1H,11H-Perfluoroundecyl methacrylate, 95%, Thermo Scientific™
CAS: 41123-44-8 Molecular Formula: C15H8F20O2 Molecular Weight (g/mol): 600.195 MDL Number: MFCD00042111 InChI Key: PUCOLQDJAFBFDR-UHFFFAOYSA-N Synonym: 1h,1h,11h-eicosafluoroundecyl methacrylate,methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl ester PubChem CID: 2776421 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCC(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F