accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41145-84-0

IUPAC Name: butyl 2-bromopropanoate
Synonyms: butyl 2-bromopropanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: INMAEXGIVZJYIJ-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C(C)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results