Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl 2-Bromopropionate 95.0+%, TCI America™
SDP

Catalog No. B103125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B103125ML Supplier TCI America Supplier No. B103125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41145-84-0
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00059469
InChI Key INMAEXGIVZJYIJ-UHFFFAOYSA-N
Synonym 2-Bromopropionic Acid Butyl Ester
PubChem CID 521024
IUPAC Name butyl 2-bromopropanoate
SMILES CCCCOC(=O)C(C)Br

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 mL
Formula Weight 209.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl 2-Bromopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.