accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClN

CAS RN 41173-36-8

IUPAC Name: 1-(4-chloro-2-nitrophenyl)pyrrolidine
Synonyms: 1-(4-chloro-2-nitrophenyl)pyrrolidine
Molecular Weight (g/mol): 226.66
Molecular Formula: C10H11ClN2O2
InChi Key: QATZMIJXHKOELL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC=C1N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results