accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41186-03-2

IUPAC Name: 1-(3-methylphenyl)piperazine
Synonyms: 1-(3-methylphenyl)piperazine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: JIWHIRLNKIUYSM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)N1CCNCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results