Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(m-Tolyl)piperazine 95.0+%, TCI America™
SDP

Catalog No. T22605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22605G 5 g
T226025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22605G Supplier TCI America Supplier No. T22605G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41186-03-2
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
InChI Key JIWHIRLNKIUYSM-UHFFFAOYSA-N
Synonym 1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine
PubChem CID 83111
IUPAC Name 1-(3-methylphenyl)piperazine
SMILES CC1=CC(=CC=C1)N2CCNCC2

Specifications

Boiling Point 156°C
Quantity 5 g
UN Number 3267
Formula Weight 176.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(m-Tolyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.