accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41191-92-8

H3COOCH3NH2

CAS RN 41191-92-8

IUPAC Name: ethyl 3-amino-4-methylbenzoate
Synonyms: ethyl 3-amino-4-methylbenzoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: MCNBNDUVWQEKNZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(N)=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results