Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Amino-4-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. E10255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10255G Supplier TCI America Supplier No. E10255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 41191-92-8
Melting Point 51°C
Color White-Yellow
Boiling Point 105°C
Molecular Formula C10H13NO2
Quantity 5 g
MDL Number MFCD01926538
Synonym 3-Amino-4-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-p-toluate, 3-Amino-p-toluic Acid Ethyl Ester
InChI Key MCNBNDUVWQEKNZ-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC(=C(C=C1)C)N
IUPAC Name ethyl 3-amino-4-methylbenzoate
Molecular Weight (g/mol) 179.219
PubChem CID 12607966
Formula Weight 179.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Amino-4-methylbenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.