accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 4122-56-9

IUPAC Name: methyl 2-formylbenzoate
Synonyms: methyl 2-formylbenzoate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: YRMODRRGEUGHTF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results