CAS RN 4122-57-0
IUPAC Name:
3-methoxy-1,3-dihydro-2-benzofuran-1-one
Synonyms:
3-methoxy-3H-2-benzofuran-1-one
Molecular Weight (g/mol):
164.16
Molecular Formula:
C9H8O3
InChi Key:
OIIJJGAFRGJQSQ-UHFFFAOYNA-N
SMILES:
COC1OC(=O)C2=CC=CC=C12
1
–
1
of
1
results
1
3-Methoxyphthalide 98.0+%, TCI America™
CAS: 4122-57-0 Molecular Formula: C9H8O3 Molecular Weight (g/mol): 164.16 InChI Key: OIIJJGAFRGJQSQ-UHFFFAOYSA-N Synonym: 3-Methoxy-1(3H)-isobenzofuranone PubChem CID: 349017 IUPAC Name: 3-methoxy-3H-2-benzofuran-1-one SMILES: COC1C2=CC=CC=C2C(=O)O1