accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4122-68-3

IUPAC Name: 2-(4-chlorophenoxy)acetyl chloride
Synonyms: 2-(4-chlorophenoxy)acetyl chloride
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: VRBVHQUSAOKVDH-UHFFFAOYSA-N
SMILES: ClC(=O)COC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results