Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenoxyacetyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C083225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C083225G 25 g
C0832100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C083225G Supplier TCI America Supplier No. C083225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4122-68-3
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00000727
InChI Key VRBVHQUSAOKVDH-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetyl chloride, p-chlorophenoxyacetyl chloride, 2-4-chlorophenoxy acetyl chloride, 4-chlorophenoxy acetyl chloride, acetyl chloride, 4-chlorophenoxy, 4-chlorophenoxyacetylchloride, acmc-1aqz7, 4-chlorophenoxyacetyl chlroride, 4chlorophenoxy acetyl chloride, vrbvhqusaokvdh-uhfffaoysa
PubChem CID 77771
IUPAC Name 2-(4-chlorophenoxy)acetyl chloride
SMILES C1=CC(=CC=C1OCC(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
UN Number 3265
Formula Weight 205.03
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenoxyacetyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.