accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHN

CAS RN 41234-43-9

IUPAC Name: ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: ethyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: RGHMPTHWVVRXHW-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CC2=CC=CC=C2CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results